Learn More
Strem, An Ascensus Company CAS# 7782-89-0. 50g. Lithium amide, 95%. MFCD00011093
SDP
Supplier:  Strem, An Ascensus Company 93030650G
Encompass
CAS# 7782-89-0. 50g. Lithium amide, 95%. MFCD00011093. Molecular Weight: 22.96. Molecular Formula: LiNH2. Color/form: off-white pwdr. Strem# 93-0306. http://www.strem.com/catalog/v/93-0306/
Catalog No. 50-901-13594