Learn More
Strem, An Ascensus Company CAS# 7550-35-8. 250g. Lithium bromide, anhydrous, 99+%. MFCD00011077
SDP
Supplier:  Strem, An Ascensus Company 930372250G
Encompass
CAS# 7550-35-8. 250g. Lithium bromide, anhydrous, 99+%. MFCD00011077. Molecular Weight: 86.85. Molecular Formula: LiBr. Color/form: white pwdr. Strem# 93-0372. http://www.strem.com/catalog/v/93-0372/
Catalog No. 50-901-13670