Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALADDIN SCIENTIFIC CORPORATION Lithium carbonate, 25g

Supplier:  ALADDIN SCIENTIFIC CORPORATION L10168425G

Encompass_Preferred

Lithium carbonate PrimorTrace 99 99% metals basisCas-554-13-2

Catalog No. 50-000-82535


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.