Learn More
Apexbio Technology LLC Lithium carbonate 554-13-2 500g
SDP
Supplier:  Apexbio Technology LLC B7705500000
Encompass_Preferred
Lithium carbonate (CAS 554-13-2) is a small-molecule inhibitor targeting inositol monophosphatase and glycogen synthase kinase-3 (GSK-3 ) It is designed to modulate intracellular signaling pathways thereby regulating processes including cell proliferation differentiation and apoptosis Lithium carbonate exerts its biological activity primarily through inhibition of inositol monophosphatase in the phosphatidylinositol signaling cascade and modulation of GSK-3 activity Based on these pharmacological properties lithium carbonate holds research potential in neuroscience psychiatric biology and cell signaling pathway studies particularly those involving inositol metabolism and GSK-3 -dependent regulation
Catalog No. 50-000-91885