Learn More
Strem, An Ascensus Company CAS# 554-13-2. 25g. Lithium carbonate (99.999%-Li) PURATREM. MFCD00011084
SDP
Supplier:  Strem, An Ascensus Company 03080025G
Encompass
CAS# 554-13-2. 25g. Lithium carbonate (99.999%-Li) PURATREM. MFCD00011084. Molecular Weight: 73.89. Molecular Formula: Li2CO3. Color/form: white pwdr. Strem# 03-0800. http://www.strem.com/catalog/v/03-0800/
Catalog No. 50-901-08619
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Accessories