Learn More
Sigma Aldrich Fine Chemicals Biosciences Lithium carbonate NISTR SR
Supplier:  Sigma Aldrich Fine Chemicals Biosciences NIST924A
Encompass
This Standard Reference Material (SRM) is intended for use as a chemical of known purity. It is intended primarily for use in the calibration and standardization of procedures and for the routine critical evaluation of the daily working standards used in these procedures. A unit of SRM 924a consists of a single glass bottle containing 30 g of the material. For more information please refer to the COA and SDS.SRM 924A cert SRM 924A SDS
Catalog No. 50-290-8393