Learn More
Sigma Aldrich Fine Chemicals Biosciences Lithium carbonate puriss.500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 62470500GF
Encompass
Lithium carbonate (Li2CO3) a lithium salt is a white monoclinic crystalline solid. Its industrial preparation has been described. In laboratory it can be synthesized by reacting carbon dioxide gas with a lithium hydroxide SOLUTIONon.
Catalog No. 50-289-8833