Learn More
Biotang Inc LITHIUM CHLORIDE 1M 50 ML, MT-180, 1 M Lithium Chloride solution (LiCl). Prepared in 18.2 megohms water and autoclaved at 15 psi for 30 min on liquid cycle. Shipped at room temperature. Store at room temperature.
Supplier:  Biotang Inc BTBB816
Encompass_Preferred
LITHIUM CHLORIDE 1M 50 ML, MT-180, 1 M Lithium Chloride solution (LiCl). Prepared in 18.2 megohms water and autoclaved at 15 psi for 30 min on liquid cycle. Shipped at room temperature. Store at room temperature.
Catalog No. 50-152-269