Learn More
Strem, An Ascensus Company CAS# 21324-40-3. 50g. Lithium hexafluorophosphate (99.9+%-Li). MFCD00011096
SDP
Supplier:  Strem, An Ascensus Company 03032550G
Encompass
CAS# 21324-40-3. 50g. Lithium hexafluorophosphate (99.9+%-Li). MFCD00011096. Molecular Weight: 151.92. Molecular Formula: LiPF6. Color/form: white xtl. Strem# 03-0325. http://www.strem.com/catalog/v/03-0325/
Catalog No. 50-901-08609