Learn More
Sigma Aldrich Fine Chemicals Biosciences Lithium perchlorate ACS re100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 205281100G
Encompass
Lithium perchlorate (LiClO4) is a colorless lithium salt. On the crystallization from its aqueous SOLUTIONon it affords lithium perchlorate trihydrate (LiClO4.3H2O). It can be synthesized by reacting lithium chloride with perchloric acid.
Catalog No. 50-289-9112