Learn More
Medchemexpress LLC Lithium sulfate 1kg
SDP
Supplier:  Medchemexpress LLC HYW0944641KG
Encompass_Preferred
Lithium sulfate is an inorganic compound that can improve the cycling life and efficiency of a 2 V/20 A H lead-acid battery Lithium sulfate can be used to study mental illnesses[1][2]
Catalog No. 50-002-05850