Learn More
Sigma Aldrich Fine Chemicals Biosciences Lithium sulfide 99.98 trac50G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 213241
Encompass
Lithium sulfide is an anti fluorite semiconductor (bandgap 3.865eV). It exists in orthorhombic and cubic structures. The densities of the orthorhombic and cubic structures are 1.75g/cm3 and 1.63g/cm3 respectively.
Catalog No. 50-290-6795