Learn More
Strem, An Ascensus Company CAS# 1907-33-1. 100g. Lithium t-butoxide, 98+%. MFCD00050479
SDP
Supplier:  Strem, An Ascensus Company 030780100G
Encompass
CAS# 1907-33-1. 100g. Lithium t-butoxide, 98+%. MFCD00050479. Molecular Weight: 80.06. Molecular Formula: LiOC4H9. Color/form: off-white pwdr. Strem# 03-0780. http://www.strem.com/catalog/v/03-0780/
Catalog No. 50-901-08618