Learn More
Sigma Aldrich Fine Chemicals Biosciences Lithium tetraborate 99.9 t1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 2225341KG
Encompass
Lithium tetraborate is a pyroelectric and piezoelectric material with potential applications in pressure sensors and surface acoustic wave (SAW) devices. In the presence of activators such as terbium and cupric ions it shows thermoluminesence property.
Catalog No. 50-289-7536