Learn More
AdipoGen Lithium triflimide
SDP
Supplier:  AdipoGen CDXL0010G050
Encompass_Preferred
Chemical. CAS 90076-65-6. Formula C2F6LiNO4S2. MW 287.09. Synthetic Used in the preparation of electrolyte suitable for lithium batteries showing very high conductivity. Used in the preparation of a chiral imidazolium salt via anion metathesis of the corresponding triflate.
Catalog No. 50-256-0618