Learn More
Strem, An Ascensus Company CAS# 2923-17-3. 100g. Lithium trifluoroacetate monohydrate, min. 97%. MFCD00149663
SDP
Supplier:  Strem, An Ascensus Company 035000100G
Encompass
CAS# 2923-17-3. 100g. Lithium trifluoroacetate monohydrate, min. 97%. MFCD00149663. Molecular Weight: 119.96 (137.97). Molecular Formula: LiOOCCF3∙H2O. Color/form: white pwdr. Strem# 03-5000. http://www.strem.com/catalog/v/03-5000/
Catalog No. 50-901-08648