Learn More
Medchemexpress LLC Lithocholic acid 3-sulfate disodium salt | 64936-81-8 | MFCD06200259 | >98.0% | 500.60 g/mol | C24H38Na2O6S | 5 MG

Supplier: Medchemexpress LLC HY113238A5MG
Lithocholic acid 3-sulfate disodium is the disodium salt of the sulfated bile acid lithocholic acid 3-sulfate. It is used in biochemical and cellular research as a GPR39 agonist to probe intracellular calcium signaling pathways and gallbladder-related biology.
- Disodium salt form, solid.
- High purity, reported ≥98%.
- Molecular weight 500.60 g/mol.
- Chemical formula C24H38Na2O6S.
- Used to study GPR39-mediated calcium signaling in cells.
- Common package size: 5 mg for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.