Learn More
Cayman Chemical LIthospermIc AcId 10mg
SDP
Supplier:  Cayman Chemical 3369410 MG
Encompass_Preferred
A polyphenol; inhibits xanthine oxidase (IC50 = 5.2 UG/ml); scavenges DPPH radicals (IC50 = 23.2 UG/ml); inhibits ROS production from PMA-stimulated isolated human neutrophils in a concentration-dependent manner; inhibits 3'-processing and 3'-joining to target DNA by HIV-1 integrase (IC50s = 0.83 and 0.48 µM, respectively); inhibits acute HIV-1 infection of H9 human lymphoma cells (IC50 = 2 µM) without inducing cytotoxicity; prevents carbon tetrachloride-induced hepatic necrosis in mice at 100 mg/kg
Catalog No. 50-294-6856