Learn More
Selleck Chemical LLC Lixisenatide 1mg 320367-13-3 Lyxumia, Adlyxin, ZP10A peptide, AVE0010
SDP
Supplier:  Selleck Chemical LLC S85171MG
Encompass_Preferred
Lixisenatide (Lyxumia, Adlyxin, ZP10A peptide, AVE0010) is a once-daily short-acting glucagon-like peptide-1 (GLP-1) receptor agonist (GLP-1RA) with an IC50 of 1.4 nM for the human GLP-1 receptor in receptor binding studies. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8259