Learn More
Apexbio Technology LLC LKB1 (AAK1 dual inhibitor), 5mg Cas No: 1093222-27-5 MFCD: MFCD15528944
SDP
Supplier:  Apexbio Technology LLC A35565
Encompass_Preferred
LKB1 is a selective inhibitor of Pim-1 kinase with Kd value of 35 nM [1].Pim-1 (proto-oncogene serine/threonine-protein kinase) is a proto-oncogene encodes by PIM1 gene and is reported to play an important role in multiple human cancers. There are other sizes available. Please inqury us for quote.
Catalog No. NC1590079