Learn More
Sino Biological Peptide Substrates: LKBtide
Supplier:  Sino Biological L1558
Encompass_Preferred
The synthetic peptide LKBtide (LSNLYHQGKFLQTFCGSPLY-RRRC) is derived from human NUAK2 (amino acid 196-215) and can be phosphorylated by protein Serine/Threonine kinase 11 (STK11), also known as LKB1.
Catalog No. 50-256-8754