Learn More
Hycult Biotech Inc LL-37 HUMAN ELISA KIT 2X96 TST
SDP
Supplier:  Hycult Biotech Inc HK32102
Encompass
The human LL-37 ELISA has been developed for the quantitative measurement of natural and recombinant LL-37 in plasma and cell culture medium.
Catalog No. NC0133159