Learn More
AnaSpec LL37 PEPTIDE 1MG
Supplier:  AnaSpec AS61302
Encompass
LL-37, Antimicrobial Peptide, human[LLGDFFRKSKEKIGKEFKRIVQRIKDFLRNLVPRTES] ; MW 4493.3; (1 mg) Antimicrobial peptide LL-37,� belonging� to the cathelicidin family, �is the first amphipathic alpha-helical peptide isolated from human.��It plays�an important role in the first line of defense against local infection and systemic invasion of pathogens at sites of inflammation and wounds.��Cytotoxic to both bacterial and normal eukaryotic cells�, LL-37�is significantly resistant to proteolytic degradation in solution.�
Catalog No. NC9896470