Learn More
ATCC LLC-MK2; Kidney; Rhesus Monkey (Macaca mulatta)
Supplier:  ATCC CCL7
Encompass
LLC-MK2 Original is a cell line exhibiting epithelial morphology that was isolated from the kidney of an adult monkey. This cell line was deposited by RN Hull. The cell has virus susceptibility to bovine viral diarrhea virus 1, human poliovirus 1, and human coxsackievirus B4.
Catalog No. 50-238-2930