Learn More
ABclonal Technology LLGL2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A1332320UL
Encompass_Preferred
Predicted to enable GTPase activator activity, PDZ domain binding activity, and myosin II binding activity. Acts upstream of or within establishment or maintenance of polarity of embryonic epithelium, labyrinthine layer development, and post-embryonic development. Predicted to be located in cytosol and intracellular membrane-bounded organelle. Predicted to be active in cortical actin cytoskeleton and plasma membrane. Is expressed in several structures, including cardiovascular system, endocrine gland, genitourinary system, gut, and nasal cavity epithelium. Orthologous to human LLGL2 (LLGL scribble cell polarity complex component 2).
Catalog No. 50-000-97996