Learn More
Medchemexpress LLC LLO (91-99) | 137856-41-8 | 99.2% | 1058.10 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP24555MG
Encompass_Preferred
LLO (91-99) is an exotoxin and a class I MHC-restricted T-cell epitope of listeriolysin (LLO). It is processed by antigen-presenting cells, binds to MHC class I molecules, and is presented to the cell surface. This product can be used in research on Listeria infections, such as listeriosis.
  • Induces cytotoxic T lymphocyte (CTL) responses.
  • Eliminates Listeria from infected cells.
  • Useful for research on Listeria infections and listeriosis.
  • Contributes to high protection efficiency against listeriosis when used with pulsed dendritic cells.
  • Promotes strong CD8+ T cell responses with IFN-γ production.
  • Helps protect against Listeria monocytogenes infection in mice.
  • Reduces liver granulomatous lesions.
Catalog No. 50-004-24470