Learn More
Medchemexpress LLC LLY-284 50mg | 2226515-75-7 | 50MG
SDP
Supplier:  Medchemexpress LLC HY107777A50MG
Encompass_Preferred
LLY-284 is the diastereomer of LLY-283 with much less active LLY-283 is a potent inhibitor of PRMT5 LLY-284 has the potential for the research of cancer diseases[1
Catalog No. 50-002-41214