Learn More
Apexbio Technology LLC LM 22A4 37988-18-4 50mg
SDP
Supplier:  Apexbio Technology LLC B565550
Encompass_Preferred
LM 22A4 (CAS 37988-18-4) is a small-molecule agonist that selectively targets the tropomyosin receptor kinase B (TrkB) a receptor implicated in neurotrophic signaling By activating TrkB-mediated pathways LM 22A4 modulates neuronal survival differentiation and synaptic plasticity This compound is extensively utilized in studies exploring neurobiological processes including neuronal maintenance and regeneration Its ability to influence TrkB signaling makes LM 22A4 a valuable tool in experimental models investigating mechanisms of neurodegeneration and neural repair
Catalog No. 50-000-88517