Learn More
Medchemexpress LLC LM11A-31 100mg | 102562-74-3 | 243.35 g/mol | C12H25N3O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY117088100MG
Encompass_Preferred
LM11A-31 a non-peptide p75NTR (neurotrophin receptor p75) modulator is an orally active and potent proNGF (nerve growth factor) antagonist LM11A-31 is an amino acid derivative with high blood-brain barrier permeability and blocks p75-mediated cell death LM11A-31 reverses cholinergic neurite dystrophy in Alzheimer s disease mouse models with mid- to late-stage disease progression[1][2]
Catalog No. 50-003-08319