Learn More
Medchemexpress LLC LM11A-31 50mg | 102562-74-3 | 243.35 g/mol | C12H25N3O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11708850MG
Encompass_Preferred
LM11A-31 is a small-molecule, non-peptide ligand of the p75 neurotrophin receptor (p75NTR) that acts as a proNGF antagonist. It is brain-penetrant and has been shown to block p75-mediated cell death and to reverse cholinergic neurite dystrophy in preclinical Alzheimer's disease models, and is supplied for research use.
  • Non-peptide p75 neurotrophin receptor ligand
  • Acts as a proNGF antagonist and blocks p75-mediated cell death
  • Brain-penetrant and orally active in preclinical models
  • Documented activity in Alzheimer's disease models
  • Provided with analytical documentation such as COA, H-NMR, and LC-MS
  • High reported purity for research applications
Catalog No. 50-002-29244