Learn More
TARGETMOL CHEMICALS INC LM11A-31 dihydrochloride 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1186150MG
Encompass_Preferred
Also available in 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. LM11A-31 dihydrochloride is a water-soluble, non-peptide with high blood-brain barrier permeability.LM11A-31 dihydrochloride, a p75NTR (neurotrophin receptor p75) Ligand, is a potent proNGF (nerve growth factor) antagonist. Purity 98%
Catalog No. 50-287-5906