Learn More
TARGETMOL CHEMICALS INC LM22A-4 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T756825MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. LM22A-4 is a brain-derived neurotrophic factor (BDNF) mimetic and agonist of the receptor tropomyosin-related kinase B (TrkB; IC50 47 nM in a fluorescence anisotropy assay) Purity ≥95%
Catalog No. 50-287-6582