Learn More
Medchemexpress LLC Lm22a-4 | 37988-18-4 | MFCD00382831 | >98.0% | 339.34 g/mol | C15H21N3O6 | 1 ML

Supplier: Medchemexpress LLC HY10067310MM 1ML
LM22A-4 is a synthetic small-molecule partial agonist of the tyrosine kinase receptor B (TrkB) used in neurobiology and neurological disease research. Supplied as a powder, it can be prepared into 10 mM stock solutions; its CAS number is 37988-18-4, molecular formula C15H21N3O6, and molecular weight 339.34 g/mol. For research use only.
- Specific agonist of tyrosine kinase receptor B (TrkB).
- Supplied as a powder for preparation of stock solutions.
- Soluble in water ≥50 mg/mL and DMSO ≥29 mg/mL.
- Includes recommended volumes for preparing 10 mM stock solutions.
- Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years).
- Intended for research use only; not for clinical or human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.