Learn More
Apexbio Technology LLC LMK 235, 10mg. Cas: 1418033-25-6 MFCD: MFCD26522892. HDAC4/HDAC5 inhibitor
SDP
Supplier:  Apexbio Technology LLC A449410
Encompass_Preferred
Description:IC50: cytotoxicity IC50 (490 nm for A2780; 320 nm for A2780 CisR); HDAC inhibition (650 nM for A2780; 320 nm for A2780 CisR)Histone deacetylases (EC 3.5.1.98, HDAC) are a class of enzymes that remove acetyl groups (O=C-CH3). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4945