Learn More
Medchemexpress LLC HY-128656 5mg Medchemexpress, LML134 CAS:1542135-76-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1286565MG
Encompass_Preferred
Medchemexpress, HY-128656 5mg LML134 CAS:1542135-76-1 LML134 (compound 18b) is an orally active and high selective Histamine 3 receptor (H3R) inverse agonist with Kis of 0.3 nM and 12 nM for hH3R cAMP and hH3R bdg. LML134 penetrates the brain rapidly, leading to high H3R occupancy, and disengages its target with a fast kinetic profile. LML134 has the potential for excessive sleep disorders. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9756