Learn More
Medchemexpress LLC Low molecular weight protein tyrosine phosphatase (LMW-PTP/ACP1), human | >95.0% | 5UG
SDP
Supplier:  Medchemexpress LLC HYP70327A5UG
Encompass_Preferred
Recombinant human low molecular weight protein tyrosine phosphatase (LMW-PTP/ACP1), supplied as a C-terminal His-tagged protein expressed in E. coli and provided lyophilized. The product is supplied with >95% purity by reducing SDS-PAGE, an approximate molecular weight of 18.0 kDa, and is formulated from 50 mM Tris-HCl, 300 mM NaCl, pH 7.4. Intended for research use; recommended reconstitution concentration ≥100 μg/mL.
  • Recombinant human sequence suitable for biochemical assays.
  • C-terminal His tag for affinity purification and detection.
  • High purity for reliable experimental results.
  • Expressed in E. coli for consistent production.
  • Lyophilized formulation for extended shelf life.
  • Provided with certificate of analysis (COA) and safety data sheet (SDS).
  • Recommended reconstitution at concentrations ≥100 μg/mL.
Catalog No. 50-001-74481