Learn More
ATCC LN-18; Glioblastoma; Human (Homo sapiens)
Supplier:  ATCC CRL2610
Encompass
LN-18 is a cell line exhibiting epithelial-like morphology that was isolated in 1976 from the right temporal lobe of a White, 65-year-old, male patient with glioblastoma. This cell line was deposited by N de Tribolet and can be used in neuroscience research.
Catalog No. 50-238-1698