Learn More
Santa Cruz Biotechnology LNCaP Cell Lysate, 500 µg/200 µl
Supplier:  Santa Cruz Biotechnology SC2231
Encompass
LNCaP Cell Lysate originates from the LNCaP cell line, a human prostate cancer cell line widely used in research to explore prostate cancer biology and oncogenesis. These cells were first derived from a metastatic lesion of human prostatic adenocarcinoma, and the lysate produced from these cells contains a comprehensive mix of proteins, DNA, RNA, and other cellular components, achieved through the lysis of LNCaP cells. This complex biochemical soup provides a snapshot of the cell's molecular environment at the time of lysis, including various cellular signaling pathways and metabolic processes. In research contexts, LNCaP Cell Lysate is primarily utilized to study the behavior of prostate cancer cells at the molecular level. It is particularly useful in proteomics research for identifying protein-protein interactions, protein expression levels, and post-translational modifications specific to prostate cancer cells.
Catalog No. NC3826496