Learn More
Glen Research Long Trebler Phosphoramidite, 0.25 g
Supplier:  Glen Research 10192502
Encompass_Preferred
Long Trebler Phosphoramidite is a special phosphoramidite building block designed for the synthesis of oligonucleotide dendrimers and complex branched DNA structures. It serves as a crucial branching reagent that enhances the properties of molecular biology applications. This phosphoramidite contributes to the stability of oligonucleotide dendrimers and improves the efficiency of PCR primer applications by making the resulting dendrimer strands resistant to degradation. It is particularly beneficial for nanotechnology applications and DNA chip technologies at higher temperatures.
  • Facilitates synthesis of oligonucleotide dendrimers
  • Increases stability and reduces exonuclease degradation
  • Enables efficient attachment of modifier amidites
  • Ideal for "bottom up" nano-assembly
  • Suitable for DNA chip technology
  • Intended for research use only
Catalog No. NC2303768