Learn More
LONGER PUMP Pumps/ Syringe pumps/ Longer Pump SP1-CX, OEM Equipment, Flow Rate: 0.0025-1250ml/min, CE Certified, Rotating valve, RS485, RS232 or CAN bus communication interface.
Supplier:  LONGER PUMP 43/67
Encompass_Preferred
Pumps/ Syringe pumps/ Longer Pump SP1-CXSpecifications- Accuracy: ≤1.0% (syringe ≥1mL)- Precision: CV≤0.05% at full stroke (with syringe ≥500μL)- Dimensions (L × W × H): 114mm × 65mm × 254mm- Weight: 2.15kg- Power supply: DC 24V, 1.5ASyringe and Drive- Syringe fitting: 1/4”-28 thread- Syringe: 50μL, 100μL, 250μL, 500μL, 1mL, 2.5mL, 5mL, 10mL, 25mL- Syringe material: Borosilicate glass, stainless steel plunger with PTFE coating, PTFE or UHMW-PE plunger tip- Resolution (standard mode): 6000 steps with 0.01mm in 1 step- Resolution (microstep mode): 48000 microsteps with 1.25μm in 1 microstep- Travel: 60mm- Time for full stroke: 1.26s-1200sValve & Valve Drive- Turn time: ≤250ms between adjacent ports- Valve options: 3-port distribution valve- Max pressure: 0.68MPA- Fitting: 1/4”-28 threade
Catalog No. NC2956826