Learn More
Cayman Chemical LipidLaunch LNP Apparent pKa Assay Kit, 96 wells, fluorometric method, uses TNS probe, for LNP development and endosomal escape evaluation
SDP
Supplier:  Cayman Chemical 70268096WELLS
Encompass_Preferred
LipidLaunch LNP Apparent pKa Assay Kit, 96 wells, fluorometric method, uses TNS probe, for LNP development and endosomal escape evaluation
Catalog No. NC2875911