Learn More
Medchemexpress LLC LNP Lipid-3 | 2648693-32-5 | C₅₀H₉₅N₃O₄ | 5 MG

Supplier: Medchemexpress LLC HY1531865MG
LNP Lipid-3 is an ionizable lipid with high purity, primarily used to synthesize lipid nanoparticles. It finds application in various drug delivery research endeavors.
- Ionizable lipid
- Used to synthesize lipid nanoparticles
- Applied in drug delivery research
- High purity (99.91%)
- Molecular weight of 802.31
- Soluble in DMSO (100 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.