Learn More
Trajan Scientific LNR INL GOOSNK AGLENT4MM 25/PK
SDP
Supplier:  Trajan Scientific 092223
Encompass_Preferred
Trajan GC Inlet Liners, Includes: Split/Splitless Liner, Diameter Inner: 4 mm, Diameter Outer: 6.3 mm, Overall Length: 78.5 mm
Catalog No. 50-253-0170