Learn More
Trajan Scientific LNR TQ CE SPLIT FOCUS 5/PK
SDP
Supplier:  Trajan Scientific 092048
Encompass_Preferred
Trajan GC Inlet Liners, Includes: Split FocusLiner; For use with 50mm needle, Diameter Inner: 5 mm, Diameter Outer: 8 mm, Overall Length: 105 mm
Catalog No. 50-255-0758