Learn More
Cayman Chemical Lnsoprazole-d4 500ug
SDP
Supplier:  Cayman Chemical 25227500 UG
Encompass_Preferred
An internal standard for the quantification of lansoprazole by GC- or LC-MS
Catalog No. 50-293-2576