Learn More
Cayman Chemical Lnsoprazole N-oxIde 25mg
SDP
Supplier:  Cayman Chemical 3542725 MG
Encompass_Preferred
A potential impurity found in bulk preparations of lansoprazole; a degradation product formed under acid or base stress
Catalog No. 50-291-3214