Learn More
Med Vet International Load/Neutral Drill Guide for 2mm plates, J0411
Supplier:  Med Vet International JOR411
Encompass_Preferred
To exert compression at a fracture site, the pilot bit must be positioned precisely in the slot in the compression plate. The load/neutral drill guide has two sleeves, one for compression (load) and one where no compression is exerted (neutral). There is one combined guide for each screw size.
Catalog No. 50-005-82635