Learn More
As One International Inc Loading Buffer Red, 5 x 1 mL
Supplier:  As One International Inc AO608104
Encompass_Preferred
5x DNA loading buffer used for loading DNA samples onto agarose or polyacrylamide gels and tracking migration during electrophoresis. Red tracking dye migrates at 300-500 bp
Catalog No. 50-165-6028