Learn More
Selleck Chemical LLC Lobaplatin S8880-1MG
SDP
Supplier:  Selleck Chemical LLC S88801MG
Encompass_Preferred
Lobaplatin (D-19466) is a third-generation platinum anti-neoplastic agent It is a platinum complex with DNA alkylating activity DMSO is not recommended to dissolve platinum-based drugs which can easily lead to drug inactivation
Catalog No. 50-000-24518