Learn More
Medchemexpress LLC Lobeglitazone (sulfate) | 763108-62-9 | MFCD28502044 | 99.6% | 578.61 g·mol⁻¹ | C24H26N4O9S2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14928A10MG
Encompass_Preferred
Lobeglitazone sulfate is a thiazolidinedione-class small molecule used for research as a peroxisome proliferator-activated receptor (PPAR) agonist. It has reported activity on PPARγ (EC50 ≈ 137.4 nM) and PPARα (EC50 ≈ 546.3 nM) and is applied in preclinical studies of metabolic, inflammatory, fibrotic, and atherosclerotic pathways. Follow the product SDS for handling and storage instructions.
  • Acts as a PPARγ and PPARα agonist with published EC50 values.
  • Supports studies of anti-diabetic, anti-inflammatory, and anti-fibrotic mechanisms.
  • High purity suitable for biochemical and cellular assays.
  • Provided as a sulfate salt to aid solubility in experimental media.
  • Stable under recommended storage; consult SDS for solvent storage conditions.
  • Supplied in small quantities appropriate for research workflows.
Catalog No. 50-002-86664